Product Details

SNP ID
rs34215645
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:4615829 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGCGCTCCTTCTCATAGCGCAG[C/T]ACCGACTGCAGAACCTCGCCCACAC
Phenotype
MIM: 604689 MIM: 609181
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AKAP3 PubMed Links

Gene Details

Gene
AKAP3
Gene Name
A-kinase anchoring protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278309.1 2929 Silent Mutation GTA,GTG V824V NP_001265238.1
NM_006422.3 2929 Silent Mutation GTA,GTG V824V NP_006413.3
XM_005253664.2 2929 Silent Mutation GTA,GTG V824V XP_005253721.1
XM_011520908.1 2929 Silent Mutation GTA,GTG V824V XP_011519210.1
XM_011520909.1 2929 Silent Mutation GTA,GTG V824V XP_011519211.1
XM_011520910.2 2929 Silent Mutation GTA,GTG V824V XP_011519212.1
Gene
DYRK4
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 4
There are no transcripts associated with this gene.

View Full Product Details