Product Details

SNP ID
rs12251874
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:24849775 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGATGCCTGGGGGGACAAACTTGA[C/T]AGCTGGCTTCCCAAGTACAGGCGTA
Phenotype
MIM: 610751
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRTFDC1 PubMed Links

Gene Details

Gene
PRTFDC1
Gene Name
phosphoribosyl transferase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282786.1 816 UTR 3 NP_001269715.1
NM_020200.6 816 UTR 3 NP_064585.1

View Full Product Details