Product Details

SNP ID
rs35014126
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:180271887 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAACGCCTGAAGAAGGATGCAG[C/G]GCGGCACCGCTGGGGGCAGTTCTAT
Phenotype
MIM: 616352 MIM: 602146
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ACBD6 PubMed Links

Gene Details

Gene
ACBD6
Gene Name
acyl-CoA binding domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032360.3 1125 Intron NP_115736.1
Gene
LHX4
Gene Name
LIM homeobox 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033343.3 1125 Missense Mutation GCG,GGG A220G NP_203129.1
XM_011510105.2 1125 Missense Mutation GCG,GGG A159G XP_011508407.1
XM_011510106.2 1125 Missense Mutation GCG,GGG A159G XP_011508408.1
XM_011510108.2 1125 Missense Mutation GCG,GGG A145G XP_011508410.1
XM_017002755.1 1125 Missense Mutation GCG,GGG A145G XP_016858244.1
Gene
LHX4-AS1
Gene Name
LHX4 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details