Product Details

SNP ID
rs11064195
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6445019 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGGGTGCAAAGAAGAGACAGCAG[C/T]GCCCAGCTTGGAGGTGCTAACTCCA
Phenotype
MIM: 186711 MIM: 607081
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD27 PubMed Links

Gene Details

Gene
CD27
Gene Name
CD27 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242.4 153 UTR 5 NP_001233.1
XM_011521042.2 153 Intron XP_011519344.1
XM_017020232.1 153 UTR 5 XP_016875721.1
XM_017020233.1 153 Intron XP_016875722.1
XM_017020234.1 153 UTR 5 XP_016875723.1
Gene
CD27-AS1
Gene Name
CD27 antisense RNA 1
There are no transcripts associated with this gene.

Gene
TAPBPL
Gene Name
TAP binding protein like
There are no transcripts associated with this gene.

View Full Product Details