Product Details

SNP ID
rs28947168
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:113006842 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGCTCATGCATTTTTATTTTTGG[C/T]TTCCTTGGGGTTGGTATATGTTATG
Phenotype
MIM: 605509
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL36A PubMed Links

Gene Details

Gene
IL36A
Gene Name
interleukin 36, alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014440.2 Intron NP_055255.1
XM_005263639.2 Intron XP_005263696.1
XM_011510965.1 Intron XP_011509267.1
XM_017003806.1 Intron XP_016859295.1

View Full Product Details