Product Details
- SNP ID
-
rs61731957
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:4274209 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAGGACATCCAACCCTACATGCGC[A/G]GAATGGTGGCCACCTGGATGCTGGA
- Phenotype
-
MIM: 123833
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCND2
PubMed Links
Gene Details
- Gene
- CCND2
- Gene Name
- cyclin D2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001759.3 |
474 |
Missense Mutation |
AGA,GGA |
R57G |
NP_001750.1 |
- Gene
- CCND2-AS1
- Gene Name
- CCND2 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details