Product Details

SNP ID
rs34057232
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30655292 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCGGCGCATCGATCTGAACTGTAC[A/G]AAGTTCTCATAGCACCACTGGGAAG
Phenotype
MIM: 615556 MIM: 603405
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATAT1 PubMed Links

Gene Details

Gene
ATAT1
Gene Name
alpha tubulin acetyltransferase 1
There are no transcripts associated with this gene.

Gene
C6orf136
Gene Name
chromosome 6 open reading frame 136
There are no transcripts associated with this gene.

Gene
DHX16
Gene Name
DEAH-box helicase 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164239.1 3126 Silent Mutation TTA,TTG L842L NP_001157711.1
NM_003587.4 3126 Silent Mutation TTA,TTG L902L NP_003578.2
XM_011514938.2 3126 Missense Mutation TTA,TTG L603L XP_011513240.1
XM_011514939.2 3126 Missense Mutation TTA,TTG L603L XP_011513241.1
XM_011514940.2 3126 Missense Mutation TTA,TTG L421L XP_011513242.1
XM_011514941.2 3126 Missense Mutation TTA,TTG L385L XP_011513243.1

View Full Product Details