Product Details
- SNP ID
-
rs35983315
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:69810513 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGAACAGGGAGGACTTCTGTGTG[C/T]GGCGGGTCTTGAAGCCCCTGCGGGG
- Phenotype
-
MIM: 164950
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FGF3
PubMed Links
Gene Details
- Gene
- FGF3
- Gene Name
- fibroblast growth factor 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005247.2 |
1003 |
Missense Mutation |
CAC,CGC |
H171R |
NP_005238.1 |
View Full Product Details