Product Details
- SNP ID
-
rs80323924
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:97661941 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAACCAAAAAACATTACACAAATAT[C/T]TGTGTATAGATGTGAGCAACCATCT
- Phenotype
-
MIM: 600469
MIM: 611153
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NCBP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75225507] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NCBP1
- Gene Name
- nuclear cap binding protein subunit 1
- Gene
- XPA
- Gene Name
- XPA, DNA damage recognition and repair factor
View Full Product Details