Product Details
- SNP ID
-
rs41279128
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:98797235 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTCGTTTACCGCTAAGGCTTGACC[A/G]CAAGCTTCCACAAATTGCCTGGAAT
- Phenotype
-
MIM: 607325
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DOCK9
PubMed Links
Gene Details
- Gene
- DOCK9
- Gene Name
- dedicator of cytokinesis 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001130048.1 |
6217 |
Silent Mutation |
TGC,TGT |
C2026C |
NP_001123520.1 |
NM_001130049.1 |
6217 |
Intron |
|
|
NP_001123521.1 |
NM_001130050.1 |
6217 |
Intron |
|
|
NP_001123522.1 |
NM_001318849.1 |
6217 |
Silent Mutation |
TGC,TGT |
C2004C |
NP_001305778.1 |
NM_015296.2 |
6217 |
Silent Mutation |
TGC,TGT |
C2027C |
NP_056111.1 |
XM_005254034.3 |
6217 |
Silent Mutation |
TGC,TGT |
C2013C |
XP_005254091.1 |
XM_005254035.3 |
6217 |
Silent Mutation |
TGC,TGT |
C1982C |
XP_005254092.1 |
XM_006719922.1 |
6217 |
Silent Mutation |
TGC,TGT |
C2061C |
XP_006719985.1 |
XM_006719924.2 |
6217 |
Silent Mutation |
TGC,TGT |
C2050C |
XP_006719987.1 |
XM_006719925.1 |
6217 |
Silent Mutation |
TGC,TGT |
C2049C |
XP_006719988.1 |
XM_006719931.3 |
6217 |
Silent Mutation |
TGC,TGT |
C2027C |
XP_006719994.1 |
XM_006719932.1 |
6217 |
Silent Mutation |
TGC,TGT |
C2026C |
XP_006719995.1 |
XM_006719936.1 |
6217 |
Silent Mutation |
TGC,TGT |
C2015C |
XP_006719999.1 |
XM_006719937.3 |
6217 |
Silent Mutation |
TGC,TGT |
C2004C |
XP_006720000.1 |
XM_017020503.1 |
6217 |
Silent Mutation |
TGC,TGT |
C2060C |
XP_016875992.1 |
XM_017020504.1 |
6217 |
Silent Mutation |
TGC,TGT |
C2046C |
XP_016875993.1 |
XM_017020505.1 |
6217 |
Silent Mutation |
TGC,TGT |
C2037C |
XP_016875994.1 |
XM_017020506.1 |
6217 |
Silent Mutation |
TGC,TGT |
C2029C |
XP_016875995.1 |
XM_017020507.1 |
6217 |
Silent Mutation |
TGC,TGT |
C2023C |
XP_016875996.1 |
XM_017020508.1 |
6217 |
Silent Mutation |
TGC,TGT |
C2015C |
XP_016875997.1 |
XM_017020509.1 |
6217 |
Silent Mutation |
TGC,TGT |
C2047C |
XP_016875998.1 |
XM_017020510.1 |
6217 |
Silent Mutation |
TGC,TGT |
C2006C |
XP_016875999.1 |
XM_017020511.1 |
6217 |
Silent Mutation |
TGC,TGT |
C2000C |
XP_016876000.1 |
XM_017020512.1 |
6217 |
Silent Mutation |
TGC,TGT |
C2035C |
XP_016876001.1 |
XM_017020513.1 |
6217 |
Silent Mutation |
TGC,TGT |
C1992C |
XP_016876002.1 |
XM_017020514.1 |
6217 |
Silent Mutation |
TGC,TGT |
C2024C |
XP_016876003.1 |
XM_017020515.1 |
6217 |
Silent Mutation |
TGC,TGT |
C2018C |
XP_016876004.1 |
XM_017020516.1 |
6217 |
Silent Mutation |
TGC,TGT |
C2012C |
XP_016876005.1 |
XM_017020517.1 |
6217 |
Silent Mutation |
TGC,TGT |
C2005C |
XP_016876006.1 |
XM_017020518.1 |
6217 |
Silent Mutation |
TGC,TGT |
C2038C |
XP_016876007.1 |
XM_017020519.1 |
6217 |
Silent Mutation |
TGC,TGT |
C1993C |
XP_016876008.1 |
XM_017020520.1 |
6217 |
Silent Mutation |
TGC,TGT |
C1981C |
XP_016876009.1 |
XM_017020521.1 |
6217 |
Silent Mutation |
TGC,TGT |
C2015C |
XP_016876010.1 |
View Full Product Details