Product Details

SNP ID
rs138830150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:75549819 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGCTGGAGGGCCGGGCACCGCGG[C/T]CATGGAGGGTCAACGCTGGCTGCCG
Phenotype
MIM: 612377 MIM: 603090
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COMMD6 PubMed Links

Gene Details

Gene
COMMD6
Gene Name
COMM domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287392.1 70 Intron NP_001274321.1
NM_001287393.1 70 Intron NP_001274322.1
NM_001287394.1 70 Intron NP_001274323.1
NM_203495.3 70 Intron NP_987091.1
NM_203497.3 70 Intron NP_987093.1
XM_011534948.2 70 Intron XP_011533250.1
Gene
UCHL3
Gene Name
ubiquitin C-terminal hydrolase L3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270952.1 70 Intron NP_001257881.1
NM_006002.4 70 UTR 5 NP_005993.1
XM_011535212.1 70 UTR 5 XP_011533514.1
XM_011535213.2 70 UTR 5 XP_011533515.1
XM_011535214.2 70 Intron XP_011533516.1
XM_017020725.1 70 UTR 5 XP_016876214.1
XM_017020726.1 70 UTR 5 XP_016876215.1
XM_017020727.1 70 UTR 5 XP_016876216.1

View Full Product Details