Product Details

SNP ID
rs34371951
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:57840938 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGAAATACTCACAGTCCTTGCTTC[A/G]CCTACTCGTCTTAAAATGACACCTT
Phenotype
MIM: 611985
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPS23 PubMed Links

Gene Details

Gene
MRPS23
Gene Name
mitochondrial ribosomal protein S23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016070.3 470 Silent Mutation GGC,GGT G136G NP_057154.2

View Full Product Details