Product Details

SNP ID
rs62640870
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:22245137 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTCACAATCAGACAAGAGCCAG[A/G]GCCCCCAGGTCCCCCCTCGCTTCTC
Phenotype
MIM: 605141
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VPREB1 PubMed Links

Gene Details

Gene
VPREB1
Gene Name
pre-B lymphocyte 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303509.1 258 Missense Mutation AGC,GGC S79G NP_001290438.1
NM_007128.3 258 Missense Mutation AGC,GGC S80G NP_009059.1

View Full Product Details