Product Details

SNP ID
hCV25613995
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67227944 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGGGGCCAATGTAAGTGCCTAC[C/T]TCCTTGCTTCTATGCCCACTTATAG
Phenotype
MIM: 606881
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FHOD1 PubMed Links

Gene Details

Gene
FHOD1
Gene Name
formin homology 2 domain containing 1
There are no transcripts associated with this gene.

Gene
LOC105369155
Gene Name
uncharacterized LOC105369155
There are no transcripts associated with this gene.

Gene
LRRC29
Gene Name
leucine rich repeat containing 29
There are no transcripts associated with this gene.

Gene
TMEM208
Gene Name
transmembrane protein 208
There are no transcripts associated with this gene.

View Full Product Details