Product Details

SNP ID
rs4253231
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:49458762 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATGATTATTAATAATAAATTAATA[A/G]TCAGAAATGCCCGTTAGAAAAAGGG
Phenotype
MIM: 609413
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ERCC6 PubMed Links

Gene Details

Gene
ERCC6
Gene Name
ERCC excision repair 6, chromatin remodeling factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000124.3 4711 UTR 3 NP_000115.1
NM_001277058.1 4711 Intron NP_001263987.1
NM_001277059.1 4711 Intron NP_001263988.1

View Full Product Details