Product Details

SNP ID
rs8190556
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:96240603 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTAGTGGCTGAGCTCCCAGGCTC[C/T]GGCTTCTCTCCACCTCGCCACATAG
Phenotype
MIM: 605573
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HSD17B3 PubMed Links

Gene Details

Gene
HSD17B3
Gene Name
hydroxysteroid 17-beta dehydrogenase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000197.1 Intron NP_000188.1
XM_011518618.2 Intron XP_011516920.1
XM_011518619.2 Intron XP_011516921.1
XM_017014671.1 Intron XP_016870160.1
XM_017014672.1 Intron XP_016870161.1
XM_017014673.1 Intron XP_016870162.1
XM_017014674.1 Intron XP_016870163.1
XM_017014675.1 Intron XP_016870164.1
XM_017014676.1 Intron XP_016870165.1
XM_017014677.1 Intron XP_016870166.1

View Full Product Details