Product Details

SNP ID
rs10798599
Assay Type
Validated
NCBI dbSNP Submissions
37
Location
Chr.1:171702323 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TACTACAAAGTTCCATGAGTGATAA[G/T]TCTGTTACCCATATCCCAGGCAGCA
Phenotype
MIM: 606909
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
VAMP4 PubMed Links

Gene Details

Gene
VAMP4
Gene Name
vesicle associated membrane protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185127.1 2988 UTR 3 NP_001172056.1
NM_003762.4 2988 UTR 3 NP_003753.2
XM_017002713.1 2988 UTR 3 XP_016858202.1

View Full Product Details