Product Details

SNP ID
rs11541583
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55387810 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACAGCTACCTGCTGGCCTGCCCA[C/G]GCACCCGCCACGGGCCCACGCTGCT
Phenotype
MIM: 603638
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIR6805 PubMed Links

Gene Details

Gene
MIR6805
Gene Name
microRNA 6805
There are no transcripts associated with this gene.

Gene
RPL28
Gene Name
ribosomal protein L28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000991.4 Intron NP_000982.2
NM_001136134.1 Intron NP_001129606.1
NM_001136135.1 Intron NP_001129607.1
NM_001136136.1 Intron NP_001129608.1
NM_001136137.1 Intron NP_001129609.1
Gene
TMEM190
Gene Name
transmembrane protein 190
There are no transcripts associated with this gene.

Gene
TMEM238
Gene Name
transmembrane protein 238
There are no transcripts associated with this gene.

View Full Product Details