Product Details

SNP ID
rs960176805
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:171185920 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAATCACAGTTACTGATGGGTCA[C/T]ATTGAGAAATTTATTAAACAACTCT
Phenotype
MIM: 603955
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FMO2 PubMed Links

Gene Details

Gene
FMO2
Gene Name
flavin containing monooxygenase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301347.1 Intron NP_001288276.1
NM_001460.4 Intron NP_001451.2

View Full Product Details