Product Details

SNP ID
rs118183800
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:60347775 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACACAGAAGTAGCCTCAACGCACA[A/G]TGAAGCTTCTCCCATGACAAAGAGA
Phenotype
MIM: 151740
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANXA2 PubMed Links

Gene Details

Gene
ANXA2
Gene Name
annexin A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002857.1 Intron NP_001002857.1
NM_001002858.2 Intron NP_001002858.1
NM_001136015.2 Intron NP_001129487.1
NM_004039.2 Intron NP_004030.1
XM_011521477.2 Intron XP_011519779.1
XM_017022090.1 Intron XP_016877579.1
XM_017022091.1 Intron XP_016877580.1
XM_017022092.1 Intron XP_016877581.1
XM_017022093.1 Intron XP_016877582.1

View Full Product Details