Product Details

SNP ID
rs72797510
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:84656861 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGCGCCGTTTCTAATGTGTCTT[C/T]TCTGTCCAGGTATACCGCTGGGCCG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHL36 PubMed Links

Gene Details

Gene
KLHL36
Gene Name
kelch like family member 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303451.1 Intron NP_001290380.1
NM_024731.3 Intron NP_079007.2
XM_005256149.2 Intron XP_005256206.1
XM_017023674.1 Intron XP_016879163.1
XM_017023675.1 Intron XP_016879164.1

View Full Product Details