Product Details

SNP ID
rs576363586
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48838968 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCACTCGCGCGTCACTATCACCCA[A/G]TTTCCTTGATGCAGAAGTAGGATCT
Phenotype
MIM: 612832 MIM: 607769
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSD17B14 PubMed Links

Gene Details

Gene
HSD17B14
Gene Name
hydroxysteroid 17-beta dehydrogenase 14
There are no transcripts associated with this gene.

Gene
PLEKHA4
Gene Name
pleckstrin homology domain containing A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161354.1 Intron NP_001154826.1
NM_020904.2 Intron NP_065955.2
XM_005259107.2 Intron XP_005259164.1
XM_006723301.2 Intron XP_006723364.1
XM_011527157.1 Intron XP_011525459.1
XM_011527158.2 Intron XP_011525460.1
XM_011527159.1 Intron XP_011525461.1
XM_011527160.1 Intron XP_011525462.1
XM_011527162.2 Intron XP_011525464.1
XM_017027037.1 Intron XP_016882526.1

View Full Product Details