Product Details

SNP ID
rs151205492
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:113553064 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACTGACATTTTTCCCCCCTAAAG[C/G]CATAGACAACAAAAGAAATTTTATT
Phenotype
MIM: 603924
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HABP2 PubMed Links

Gene Details

Gene
HABP2
Gene Name
hyaluronan binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177660.2 87 Intron NP_001171131.1
NM_004132.4 87 UTR 5 NP_004123.1

View Full Product Details