Product Details

SNP ID
rs78163411
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:120851534 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGACGCCGAGCTTCCAGGTCGCC[A/G]GGATCGGCCAGGAATGTGTGAGGGG
Phenotype
MIM: 606417
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WDR11 PubMed Links

Gene Details

Gene
WDR11
Gene Name
WD repeat domain 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018117.11 Intron NP_060587.8
XM_005269962.2 Intron XP_005270019.1
XM_005269963.2 Intron XP_005270020.1
XM_005269964.2 Intron XP_005270021.1
XM_017016397.1 Intron XP_016871886.1
XM_017016398.1 Intron XP_016871887.1
XM_017016399.1 Intron XP_016871888.1
XM_017016400.1 Intron XP_016871889.1
Gene
WDR11-AS1
Gene Name
WDR11 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details