Product Details

SNP ID
rs41287948
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:119892754 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTGGTGTGGTACCGGGTACCCGG[A/G]GACGTGTATCGGACGGTGGGCCGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEC23IP PubMed Links

Gene Details

Gene
SEC23IP
Gene Name
SEC23 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007190.3 25 UTR 5 NP_009121.1
XM_005269469.2 25 UTR 5 XP_005269526.1
XM_005269470.2 25 UTR 5 XP_005269527.1
XM_017015610.1 25 UTR 5 XP_016871099.1

View Full Product Details