Product Details

SNP ID
rs7094098
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:119898593 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAACTGCAGCAACCTCAGTTGGACA[A/G]TCAGGATTCCCCAAGCCCCTGACTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEC23IP PubMed Links

Gene Details

Gene
SEC23IP
Gene Name
SEC23 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007190.3 566 Silent Mutation CAA,CAG Q110Q NP_009121.1
XM_005269469.2 566 Silent Mutation CAA,CAG Q110Q XP_005269526.1
XM_005269470.2 566 Silent Mutation CAA,CAG Q63Q XP_005269527.1
XM_017015610.1 566 Silent Mutation CAA,CAG Q63Q XP_016871099.1

View Full Product Details