Product Details

SNP ID
rs34816476
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31589604 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATAGAATCCAGCCCGAAGGAGGA[A/G]GACTGTACCAGCCCCTAGCTGAGGA
Phenotype
MIM: 109170 MIM: 600978 MIM: 611550
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LST1 PubMed Links

Gene Details

Gene
LST1
Gene Name
leukocyte specific transcript 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166538.1 628 Intron NP_001160010.1
NM_007161.3 628 Intron NP_009092.3
NM_205837.2 628 Intron NP_995309.2
NM_205838.2 628 Intron NP_995310.2
NM_205839.2 628 Intron NP_995311.2
NM_205840.2 628 Intron NP_995312.2
XM_006715206.3 628 Intron XP_006715269.1
XM_006715209.3 628 Intron XP_006715272.1
XM_006715210.3 628 Intron XP_006715273.1
XM_011514914.2 628 Intron XP_011513216.1
Gene
LTB
Gene Name
lymphotoxin beta
There are no transcripts associated with this gene.

Gene
NCR3
Gene Name
natural cytotoxicity triggering receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145466.1 628 Missense Mutation CTC,TTC L140F NP_001138938.1
NM_001145467.1 628 Missense Mutation CTC,TTC L140F NP_001138939.1
NM_147130.2 628 Missense Mutation CTC,TTC L140F NP_667341.1
XM_006715049.3 628 Missense Mutation CTC,TTC L140F XP_006715112.1
XM_011514459.2 628 Missense Mutation CTC,TTC L115F XP_011512761.1

View Full Product Details