Product Details

SNP ID
hCV25633536
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:106775017 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCATGCATCCTGCATCCCACTA[C/T]CCCCACTGTAGCGCCCACTCCTCTG
Phenotype
MIM: 609147
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100287944 PubMed Links

Gene Details

Gene
LOC100287944
Gene Name
uncharacterized LOC100287944
There are no transcripts associated with this gene.

Gene
RIC8B
Gene Name
RIC8 guanine nucleotide exchange factor B
There are no transcripts associated with this gene.

View Full Product Details