Product Details
- SNP ID
-
rs6757411
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:117819599 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CTTCTGTTTGTTGATCTCATTTGGA[A/T]TTCTATTTTAAGGAAAAATTTTTTC
- Phenotype
-
MIM: 606355
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DDX18
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139417430] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DDX18
- Gene Name
- DEAD-box helicase 18
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006773.3 |
|
Intron |
|
|
NP_006764.3 |
View Full Product Details