Product Details
- SNP ID
-
rs35525096
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:18086965 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGCACTGTTTTCTGTTCTTGAAGAA[C/T]GATGGGTGGATCATCAGACTGGCAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SAAL1
PubMed Links
Gene Details
- Gene
- SAAL1
- Gene Name
- serum amyloid A like 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_138421.2 |
993 |
Missense Mutation |
ATT,GTT |
I315V |
NP_612430.2 |
View Full Product Details