Product Details

SNP ID
rs10020783
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:74054393 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACGCAGCGCAGTTCAGTGTACAAGT[A/C]ACTGTCTACACTTACAGCTGGGGTA
Phenotype
MIM: 611591
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PPBPP2 PubMed Links

Gene Details

Gene
PPBPP2
Gene Name
pro-platelet basic protein pseudogene 2
There are no transcripts associated with this gene.

View Full Product Details