Product Details

SNP ID
rs3739337
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:66429842 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCGCCTCTGTGGGGCGCTTTCAGGA[A/G]CGCCTCCCCAAGGAGAAGGTGCCCC
Phenotype
MIM: 611083
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADHFE1 PubMed Links

Gene Details

Gene
ADHFE1
Gene Name
alcohol dehydrogenase, iron containing 1
There are no transcripts associated with this gene.

Gene
RRS1
Gene Name
ribosome biogenesis regulator homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015169.3 815 Silent Mutation GAA,GAG E237E NP_055984.1
Gene
RRS1-AS1
Gene Name
RRS1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details