Product Details

SNP ID
rs78873228
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:99205119 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGAGGCTTCATTATAAAGGAAA[A/G]GTCATTCAAAGTAACTGCTGAAGTT
Phenotype
MIM: 103735
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADH6 PubMed Links

Gene Details

Gene
ADH6
Gene Name
alcohol dehydrogenase 6 (class V)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000672.3 Intron NP_000663.1
NM_001102470.1 Intron NP_001095940.1
Gene
LOC100507053
Gene Name
uncharacterized LOC100507053
There are no transcripts associated with this gene.

View Full Product Details