Product Details

SNP ID
rs59092606
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.19:15618122 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
TCAACTTGTGCCACCCTGACATCGT[C/T]CGATCTGTCATCAATACCTCAGGTA
Phenotype
MIM: 611545
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYP4F8 PubMed Links

Gene Details

Gene
CYP4F8
Gene Name
cytochrome P450 family 4 subfamily F member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007253.3 322 Silent Mutation GTC,GTT V107V NP_009184.1
XM_017026232.1 322 Silent Mutation GTC,GTT V107V XP_016881721.1

View Full Product Details