Product Details

SNP ID
rs35701751
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89577589 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCCTGCATCCCGTGTTCTCCAA[A/G]GTCTTCACGGTGGACTACTACTTCG
Phenotype
MIM: 605689
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CPNE7 PubMed Links

Gene Details

Gene
CPNE7
Gene Name
copine 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014427.4 30 Silent Mutation AAA,AAG K75K NP_055242.1
NM_153636.2 30 Silent Mutation AAA,AAG K75K NP_705900.1
XM_011523000.1 30 Silent Mutation AAA,AAG K75K XP_011521302.1
XM_011523001.2 30 UTR 5 XP_011521303.1
XM_017023138.1 30 Silent Mutation AAA,AAG K75K XP_016878627.1
XM_017023139.1 30 Silent Mutation AAA,AAG K79K XP_016878628.1
XM_017023140.1 30 Silent Mutation AAA,AAG K79K XP_016878629.1
XM_017023141.1 30 Intron XP_016878630.1

View Full Product Details