Product Details
- SNP ID
-
rs35541730
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
34
- Location
-
Chr.1:19814480 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGCAGAAGAGAGTCCTGGAGGAGG[G/T]CCAGCAGCTGCCCCGGGTGCTGGGG
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RNF186
PubMed Links
Gene Details
- Gene
- RNF186
- Gene Name
- ring finger protein 186
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_019062.1 |
799 |
Missense Mutation |
ACC,CCC |
T208P |
NP_061935.1 |
View Full Product Details