Product Details
- SNP ID
-
hCV25640760
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:141303426 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGACCAAATTGTATTATGGCTTTTT[A/G]CTATCTTCCCTGACATCTCCATTTC
- Phenotype
-
MIM: 608157
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SLC25A2
PubMed Links
Gene Details
- Gene
- SLC25A2
- Gene Name
- solute carrier family 25 member 2
There are no transcripts associated with this gene.
View Full Product Details