Product Details

SNP ID
rs143436570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:2405509 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCATGCCCACCCTCATCCCACG[C/T]TCCCCAAACAGGCTTACTCCGGGGG
Phenotype
MIM: 186845 MIM: 604600 MIM: 603852
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD81 PubMed Links

Gene Details

Gene
CD81
Gene Name
CD81 molecule
There are no transcripts associated with this gene.

Gene
TRPM5
Gene Name
transient receptor potential cation channel subfamily M member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014555.3 Intron NP_055370.1
XM_017017628.1 Intron XP_016873117.1
Gene
TSSC4
Gene Name
tumor suppressing subtransferable candidate 4
There are no transcripts associated with this gene.

View Full Product Details