Product Details
- SNP ID
-
rs35157623
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:32575535 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGTCCAGGTGCCAGGCAGCCCAGG[G/T]CGAGTGGTGCCCACAGGCTTGGGCT
- Phenotype
-
MIM: 608365
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TCP10L
PubMed Links
Gene Details
- Gene
- TCP10L
- Gene Name
- t-complex 10-like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_144659.5 |
2003 |
UTR 3 |
|
|
NP_653260.1 |
View Full Product Details