Product Details

SNP ID
rs61737278
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:124451194 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGTCCCTGTATGCTCACGCAGCT[C/G]CCGGATCCTGTTCCTTCGAAGAGGG
Phenotype
MIM: 611244
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
TRMT12 PubMed Links

Gene Details

Gene
TRMT12
Gene Name
tRNA methyltransferase 12 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017956.3 388 Silent Mutation CTC,CTG L89L NP_060426.2

View Full Product Details