Product Details

SNP ID
rs34025088
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:178514734 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAAAGCAGTTAAACAAGAAGGGC[A/G]GTTTACCAAAGCAGGAGTGACACAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TEX35 PubMed Links

Gene Details

Gene
TEX35
Gene Name
testis expressed 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170722.1 237 Missense Mutation CAG,CGG Q50R NP_001164193.1
NM_001170723.1 237 Missense Mutation CAG,CGG Q42R NP_001164194.1
NM_001170724.1 237 Missense Mutation CAG,CGG Q42R NP_001164195.1
NM_032126.4 237 Missense Mutation CAG,CGG Q42R NP_115502.2

View Full Product Details