Product Details

SNP ID
rs16891235
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:26017314 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTTTGCTAGCCCCCGTGGCCTTT[C/T]TGAGCTTTTTAGATGCACCCGTTGC
Phenotype
MIM: 142709 MIM: 602810 MIM: 602822 MIM: 602829
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HIST1H1A PubMed Links

Gene Details

Gene
HIST1H1A
Gene Name
histone cluster 1, H1a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005325.3 499 Missense Mutation AAA,AGA K140R NP_005316.1
Gene
HIST1H3A
Gene Name
histone cluster 1, H3a
There are no transcripts associated with this gene.

Gene
HIST1H4A
Gene Name
histone cluster 1, H4a
There are no transcripts associated with this gene.

Gene
HIST1H4B
Gene Name
histone cluster 1, H4b
There are no transcripts associated with this gene.

View Full Product Details