Product Details

SNP ID
rs62000994
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5301394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTAAACGTGATATCAGCACAGG[C/T]GAGTTTTATGACATCTTGATGGAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR51B4 PubMed Links

Gene Details

Gene
OR51B4
Gene Name
olfactory receptor family 51 subfamily B member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033179.2 553 Missense Mutation ACC,GCC T185A NP_149419.2

View Full Product Details