Product Details

SNP ID
hCV25645608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:155916224 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGAAGGTTAATATACTAATATAA[C/G]AAAGGAAAATTCAAGCACTGTGTGG
Phenotype
MIM: 609591
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KIAA0907 PubMed Links

Gene Details

Gene
KIAA0907
Gene Name
KIAA0907
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014949.3 Intron NP_055764.2
Gene
RIT1
Gene Name
Ras like without CAAX 1
There are no transcripts associated with this gene.

Gene
SCARNA4
Gene Name
small Cajal body-specific RNA 4
There are no transcripts associated with this gene.

Gene
SNORA80E
Gene Name
small nucleolar RNA, H/ACA box 80E
There are no transcripts associated with this gene.

View Full Product Details