Product Details
- SNP ID
-
rs9665861
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:55935485 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAAGAATCTACCTTTGATCTTAGAA[C/T]TTTCTCAGCTGCATCCTTTACATCT
- Phenotype
-
MIM: 608496
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR5I1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs61995929,rs9665863] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR5I1
- Gene Name
- olfactory receptor family 5 subfamily I member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006637.1 |
916 |
Missense Mutation |
ATT,GTT |
I306V |
NP_006628.1 |
View Full Product Details