Product Details

SNP ID
rs116922372
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:102692887 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGTTTTTGTGCTGTCTTTTTTCA[C/T]AGTCAACACAGTCTCTCAAGTATCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MMP27 PubMed Links

Gene Details

Gene
MMP27
Gene Name
matrix metallopeptidase 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022122.2 Intron NP_071405.2
XM_011542948.2 Intron XP_011541250.1
XM_011542949.2 Intron XP_011541251.1
XM_011542950.2 Intron XP_011541252.1
XM_017018120.1 Intron XP_016873609.1

View Full Product Details