Product Details

SNP ID
rs752813663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:57100743 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGTCCAGAAAGGATAGTTTCTCA[A/G]ACTTCCTCTGTTGAGAACCCATTGG
Phenotype
MIM: 615708
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF451 PubMed Links

Gene Details

Gene
ZNF451
Gene Name
zinc finger protein 451
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031623.2 328 Intron NP_001026794.1
NM_001257273.1 328 Silent Mutation CAA,CAG Q109Q NP_001244202.1
NM_015555.2 328 Intron NP_056370.2
XM_005248994.1 328 Intron XP_005249051.1
XM_011514460.1 328 Intron XP_011512762.1
XM_011514461.1 328 Intron XP_011512763.1
XM_011514462.2 328 Intron XP_011512764.1
XM_011514463.2 328 Silent Mutation CAA,CAG Q81Q XP_011512765.1

View Full Product Details