Product Details

SNP ID
rs193115427
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:26053716 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTATGAATAGTTTGGGGTTTTTT[C/T]CTTTTCTAAGGGTAAAGAGAACAGG
Phenotype
MIM: 609617 MIM: 606131
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC30A2 PubMed Links

Gene Details

Gene
SLC30A2
Gene Name
solute carrier family 30 member 2
There are no transcripts associated with this gene.

Gene
TRIM63
Gene Name
tripartite motif containing 63
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032588.3 1368 Intron NP_115977.2
XM_017002559.1 1368 UTR 3 XP_016858048.1

View Full Product Details