Product Details

SNP ID
rs61736224
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:88678611 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAAAGGCCCTGCTGCAGTGCGAG[A/C]AGGCATAGGGCTTCTCCCCTGTGGG
Phenotype
MIM: 612741
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SNAI3 PubMed Links

Gene Details

Gene
SNAI3
Gene Name
snail family transcriptional repressor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178310.3 784 Missense Mutation TGC,TTC C239F NP_840101.1
Gene
SNAI3-AS1
Gene Name
SNAI3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details