Product Details
- SNP ID
-
rs61737512
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:153182743 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGGTTTTCTGCTCCTCAAATATCG[T/A]GCCAGGGAGCCAGTCACAAAGGCAG
- Phenotype
-
MIM: 300016
- Polymorphism
- T/A, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MAGEA1
PubMed Links
Gene Details
- Gene
- MAGEA1
- Gene Name
- MAGE family member A1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004988.4 |
574 |
Silent Mutation |
CGA,CGT |
R118R |
NP_004979.3 |
View Full Product Details